ContactUsLinkComponent
Powder cell holder set for dra-ca-3 for cary 1/3. includes a pre-packed ptfe powder cell for use as a reflectance standard and powde r cell holder. 1 * 1 items
Catalog # VARI7910037000
Supplier:  VARIAN
Powder cell holder set for dra-ca-3 for cary 1/3. includes a pre-packed ptfe powder cell for use as a reflectance standard and powde r cell holder. 1 * 1 items
Catalog # VARI7910037000
Supplier:  VARIAN
Frequently Bought Together