ContactUsLinkComponent
Crosslab performance maintenance kit, for ultimate 3 hpg-32p pump, contains 1 stirrer, 1 silicone tubing (2.8mm od x 1.3mm id x 1.5m l), 1 pharmed tubing for peristaltic pump (3.2mm od x 1.6mm id x 18mm l), 1 fep tubing (4.5mm od x 3.mm id x 2m l), 1 * 1 items
Catalog # HEWL8002-0905
Supplier:  Agilent
Crosslab performance maintenance kit, for ultimate 3 hpg-32p pump, contains 1 stirrer, 1 silicone tubing (2.8mm od x 1.3mm id x 1.5m l), 1 pharmed tubing for peristaltic pump (3.2mm od x 1.6mm id x 18mm l), 1 fep tubing (4.5mm od x 3.mm id x 2m l), 1 * 1 items
Catalog # HEWL8002-0905
Supplier:  Agilent
Frequently Bought Together