ContactUsLinkComponent
Ketones sis kit, radial icp w/cycl. s/c. for use with ketone based solvents. incl double pass glass cyclonic s/chamber, conikal neb, torch (1.4mm id inj.) + all necessary marprene/solvent tubing 1 * 1 items
Catalog # VARI9910127200
Supplier:  VARIAN
Ketones sis kit, radial icp w/cycl. s/c. for use with ketone based solvents. incl double pass glass cyclonic s/chamber, conikal neb, torch (1.4mm id inj.) + all necessary marprene/solvent tubing 1 * 1 items
Catalog # VARI9910127200
Supplier:  VARIAN
Frequently Bought Together