ContactUsLinkComponent
Aromatics sis kit, axial icp w/inert s/c. for use with aromatic solvents. incl v-groove neb, inert sturman-masters s/chamber, torch (1.4 mm id injector) + all necessary viton/solvent tubing 1 * 1 items
  VARI9910126900
 :  VARIAN
Aromatics sis kit, axial icp w/inert s/c. for use with aromatic solvents. incl v-groove neb, inert sturman-masters s/chamber, torch (1.4 mm id injector) + all necessary viton/solvent tubing 1 * 1 items
  VARI9910126900
 :  VARIAN
Frequently Bought Together