ContactUsLinkComponent
Ketones sis kit, axial icp w/inert s/c. for use with ketone based solvents. incl v-groove neb, inert sturman-masters s/chamber, torc h (1.4 mm id inj.) + all necessary marprene/solvent tubing 1 * 1 items
Catalog # VARI9910127300
Supplier:  VARIAN
Ketones sis kit, axial icp w/inert s/c. for use with ketone based solvents. incl v-groove neb, inert sturman-masters s/chamber, torc h (1.4 mm id inj.) + all necessary marprene/solvent tubing 1 * 1 items
Catalog # VARI9910127300
Supplier:  VARIAN
Frequently Bought Together