ContactUsLinkComponent
Snap vial and cap convenience pk 5 contains 5 clear glass snap top 2ml vials w/write-on spot,5 clear pe caps with ptfe/red rubber se pta and a reusable 6-drawer cabinet 1 * 500 items
Catalog # HEWL5190-2240
Supplier:  Agilent
Snap vial and cap convenience pk 5 contains 5 clear glass snap top 2ml vials w/write-on spot,5 clear pe caps with ptfe/red rubber se pta and a reusable 6-drawer cabinet 1 * 500 items
Catalog # HEWL5190-2240
Supplier:  Agilent
Frequently Bought Together