ContactUsLinkComponent
.45um rc mini-uniprep, 1/pack regenerated cellulose for aquaeous or organic solvents, very low non-specific protein binding membrane 1 * 100 items
Catalog # HEWL5190-1418
Supplier:  Agilent
.45um rc mini-uniprep, 1/pack regenerated cellulose for aquaeous or organic solvents, very low non-specific protein binding membrane 1 * 100 items
Catalog # HEWL5190-1418
Supplier:  Agilent
Frequently Bought Together