ContactUsLinkComponent
Flat flange lid, LF 200, center neck NS 45/40, 1 side neck 45° KF 50, 1 side neck 45° NS 29/32 and 1 side neck straigth NS 14/23, DU RAN® 1 * 1 items
Catalog # LENZ06104570
Flat flange lid, LF 200, center neck NS 45/40, 1 side neck 45° KF 50, 1 side neck 45° NS 29/32 and 1 side neck straigth NS 14/23, DU RAN® 1 * 1 items
Catalog # LENZ06104570
Frequently Bought Together