ContactUsLinkComponent
Vacuum cleaner AVSD-20/25L DT #111388 HEPA Filter - Included with an efficiency of 99.995% on 0.3 micron. Tested: IEST RP-CC001.3. H14 by MPPS method as per EN 1822. Pneumatically (air) operated Available with Manual Filter Shaker (MFS) For dry recov 1 * 1 Each
Catalog # TIGV111388
Supplier:  TIGER-VAC
Vacuum cleaner AVSD-20/25L DT #111388 HEPA Filter - Included with an efficiency of 99.995% on 0.3 micron. Tested: IEST RP-CC001.3. H14 by MPPS method as per EN 1822. Pneumatically (air) operated Available with Manual Filter Shaker (MFS) For dry recov 1 * 1 Each
Catalog # TIGV111388
Supplier:  TIGER-VAC
Frequently Bought Together