ContactUsLinkComponent
Poly tubing #LT104:3600 100µ 36´ x 500´ x 004 Ultraclean Clear poly tubing Meets NASA JPAG 5322.1 level 100 Double barrier packed 1 * 1 items
Catalog # KNFCLT103600
Supplier:  KNF CLEANROOM
Poly tubing #LT104:3600 100µ 36´ x 500´ x 004 Ultraclean Clear poly tubing Meets NASA JPAG 5322.1 level 100 Double barrier packed 1 * 1 items
Catalog # KNFCLT103600
Supplier:  KNF CLEANROOM
Frequently Bought Together