Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Mask carriers

Mask carriers

Pocket spacing: 3,2 mm, pocket flat: 1,6 mm, 25 masks, PP, Blue, 107x36x28 mm
Catalog # ENTEA66-0603
Supplier:   ENTEGRIS
Mask carriers
Catalog # ENTEA66-0603
Supplier:  ENTEGRIS
Mask carriers
Mask carriers

Pocket spacing: 3,2 mm, pocket flat: 1,6 mm, 25 masks, PP, Blue, 107x36x28 mm

Price
Inquire For Price
Log in for availability
1 * 4 KS
Product Details & Documents

Miscellaneous mask carriers.

Specifications
  • For:
    25 masks
  • Colour:
    Blue
  • Description:
    Pocket spacing: 3,2 mm, pocket flat: 1,6 mm
  • Material:
    PP
  • Seq:
    03
  • W×D×H:
    107x36x28 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.