ContactUsLinkComponent
Polypropylene-coverall white / CoverCle 100% Polypropylene, white, 55g/sqm, anti-static, CE Category 1,zipper in front, with hood single packed 1 * 50 items
Catalog # ASATCC-XL
Dodavatel:  ASATEX
Polypropylene-coverall white / CoverCle 100% Polypropylene, white, 55g/sqm, anti-static, CE Category 1,zipper in front, with hood single packed 1 * 50 items
Catalog # ASATCC-XL
Dodavatel:  ASATEX
Frequently Bought Together