Orders LinkContactUsLinkComponent
Mask carriers
40 masks, PFA, Natural, 150x107x46 mm
  ENTEA19-01-0215
 :   ENTEGRIS
Mask carriers
  ENTEA19-01-0215
 :  ENTEGRIS
Mask carriers
Mask carriers
40 masks, PFA, Natural, 150x107x46 mm
1 * 4 KS
Miscellaneous mask carriers.
  • For:
    40 masks
  • Colour:
    Natural
  • Material:
    PFA
  • Seq:
    01
  • W×D×H:
    150x107x46 mm