ContactUsLinkComponent
LDPE Tubing antistatic pink 150mm x 100 m Roll antistatic, definition: ultra clean, antistatic pink PE tubing Cleanliness level: 200 (according to IEST-STD-CC1246D) ASRE00-0231-0001 1 * 1 Rolle
Catalog # ATMIASRE0002310001
Supplier:  PALL LIFESCIENCES
LDPE Tubing antistatic pink 150mm x 100 m Roll antistatic, definition: ultra clean, antistatic pink PE tubing Cleanliness level: 200 (according to IEST-STD-CC1246D) ASRE00-0231-0001 1 * 1 Rolle
Catalog # ATMIASRE0002310001
Supplier:  PALL LIFESCIENCES
Frequently Bought Together