Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Microincinerators, e-Loop

Microincinerators, e-Loop

e-Loop with 15 mm tube diameter, 710-1471
Catalog # 710-1471
Supplier:   BioTool
Microincinerators, e-Loop
Catalog # 710-1471
Supplier:  BioTool
Microincinerators, e-Loop
Microincinerators, e-Loop

e-Loop with 15 mm tube diameter, 710-1471

Price
19 180,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents

Electrical sterilisation systems which are suitable for sterilising loops, needles and instruments. Ideal for use in laboratories where using gases or naked flames is not permitted. The e-Loop has a wear resistant quartz glass tube which is protected by a perforated stainless steel outer design. Choice of two units with different tube diameters, the larger is ideal for sterilising small instruments such as forceps and scissors. The heating system provides uniform heat. The opening eliminates the potential aerosol of pathogenic microorganisms.

  • Loops and needles sterilised in 5 to 7 s
  • Integral spray protector prevents hazardous gases, flames and infectious splatter harming user
  • Sterilisation tube has self-locking angular adjustment, there are seven different positions
  • Tray for three instruments
  • Residual heat warning to prevent burns

 : Optional accessories include a holder specifically for BioTool inoculation loop holders, a stainless steel tray which is positioned on the left hand side of the steriliser and stainless steel inoculation loop holders for loops with wire Ø 0,6 to 1 mm.

Specifications
  • Model:
    710-1471
  • Description:
    e-Loop with 15 mm tube diameter
  • Seq:
    01
  • Cat. No.:
    710-1471
  • W×D×H:
    175×139×129 mm
  • Temperature range:
    900 - 950 °C
  • Power supply:
    120 W
  • Ø×L:
    15×118 mm
  • Weight:
    1,1 kg
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stainless steel tray for positioning on left hand side
    Stainless steel tray for positioning on left hand side
    710-1575
    Log in for availability
    1 * 1 KS - 1 540,00 Kč

Product Information

  • Catalog #
    Availability
    Price
  • Holding device for BioTool loop holders
    Holding device for BioTool loop holders
    710-1473
    Log in for availability
    1 * 1 KS - 1 360,00 Kč

Product Information

  • Catalog #
    Availability
    Price
  • Quartz glass heating element for e-Loop 230 V
    Quartz glass heating element for e-Loop 230 V
    710-1573
    Log in for availability
    1 * 1 KS - 8 380,00 Kč

Product Information

  • Catalog #
    Availability
    Price
  • Spare ceramic annealing tube for e-Loop standard, all voltages
    Spare ceramic annealing tube for e-Loop standard, all voltages
    710-1574
    Log in for availability
    1 * 1 KS - 2 230,00 Kč