Propidium Iodide
Glass bottle for solids, 25 mg
Catalog # CAYM14289-25
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
- CAS Number:25535-16-4
- Formula:C₂₇H₃₄I₂N₄
- MDL Number:MFCD00011921
- Molecular Weight:668.4 g/mol
- EINECS:247-081-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg



