Biotix Plate Seal Tape, 100/case. 2mm thick, non-pierceable polypropeylene film, functional temperatrure range -40C-to 125 C. Not reccomended for sealing raised rim plates. 1 * 100 items
Catalog # 732-2488
Supplier: BIOTIX
Frequently Bought Together