ContactUsLinkComponent
Precision heater block thermostat for 12 SR reaction vessels, 20 ... 399 °C 1 * 1 items
Catalog # 717-1012
Supplier:  BEHR
Precision heater block thermostat for 12 SR reaction vessels, 20 ... 399 °C 1 * 1 items
Catalog # 717-1012
Supplier:  BEHR
Frequently Bought Together