0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Ultra Protein hydrolysate Chemical kit. Contains 2 Sodium Citrate Buffer pH 3.2 2 litres, 2 Sodium Citrate Buffer pH 4.25 2 litres, 1 Sodium Citrate Buffer pH 6.45 2 1 * 1 items
WAPR80-2117-78
:
WALDEN PRECISION APPARATUS
Ultra Protein hydrolysate Chemical kit. Contains 2 Sodium Citrate Buffer pH 3.2 2 litres, 2 Sodium Citrate Buffer pH 4.25 2 litres, 1 Sodium Citrate Buffer pH 6.45 2 1 * 1 items
WAPR80-2117-78
:
WALDEN PRECISION APPARATUS
Frequently Bought Together
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals