ContactUsLinkComponent
Biotix 1250ul LEER pipette tip racks for Tip Eject Reload System. LEER racks come 10 Trays/Pack, 5 Packs/Case. These racks are comp atible with 1250ul Tip Eject pipette tips. 1 * 10 items
Catalog # 613-2825
Supplier:  BIOTIX
Biotix 1250ul LEER pipette tip racks for Tip Eject Reload System. LEER racks come 10 Trays/Pack, 5 Packs/Case. These racks are comp atible with 1250ul Tip Eject pipette tips. 1 * 10 items
Catalog # 613-2825
Supplier:  BIOTIX
Frequently Bought Together