ContactUsLinkComponent
Biotix 10 µL Extended Length Reload System Pipette Tips. Tip Eject Reload System 96 Tip/Card, 20 Cards/Pack, 5 Pack/Case. Patented features: FlexFit™ provides universal fit and superior seal, X-Resin™ for low sample retention, Blade™ for accuracy, s 1 * 1.920 items
Catalog # 613-2819
Supplier:  BIOTIX
Biotix 10 µL Extended Length Reload System Pipette Tips. Tip Eject Reload System 96 Tip/Card, 20 Cards/Pack, 5 Pack/Case. Patented features: FlexFit™ provides universal fit and superior seal, X-Resin™ for low sample retention, Blade™ for accuracy, s 1 * 1.920 items
Catalog # 613-2819
Supplier:  BIOTIX
Frequently Bought Together