ContactUsLinkComponent
pH combination electrode with Memosens screw plug head (ATEX II 1/G) low maintenance with NTC 30K, glass shaft, 3 x ceramic junction , gel electrolyte, Silamid-reference system, sphere membrane, A-glass, length 225 mm, 12 mm Ø, -5...+80 °C, 0...14 pH 1 * 1 items
Catalog # SCOI285114770
Supplier:  SI Analytics
pH combination electrode with Memosens screw plug head (ATEX II 1/G) low maintenance with NTC 30K, glass shaft, 3 x ceramic junction , gel electrolyte, Silamid-reference system, sphere membrane, A-glass, length 225 mm, 12 mm Ø, -5...+80 °C, 0...14 pH 1 * 1 items
Catalog # SCOI285114770
Supplier:  SI Analytics
Frequently Bought Together