ContactUsLinkComponent
Metall-glassreference-electrode with plug head Glass shaft, sulfidized silver sensor, A membrane glass as reference, length 120 mm, 12 mm Ø, -5...+100 °C, 0...14 pH 1 * 1 items
Katalogové číslo SCOI285102110
Supplier:  SI Analytics
Metall-glassreference-electrode with plug head Glass shaft, sulfidized silver sensor, A membrane glass as reference, length 120 mm, 12 mm Ø, -5...+100 °C, 0...14 pH 1 * 1 items
Katalogové číslo SCOI285102110
Supplier:  SI Analytics
Frequently Bought Together