ContactUsLinkComponent
pH combination electrode with Memosens screw plug head with Liquid electrolyt, NTC 30K, plastic adapter for hose connection incl. Pg 13.5 thread, platinum junction, electrolyte L 300, sphere membrane, S-glass, length 120 mm, 12 mm Ø, +10...+135°C, 0. 1 * 1 items
Catalog # SCOI285118200
Supplier:  SI Analytics
pH combination electrode with Memosens screw plug head with Liquid electrolyt, NTC 30K, plastic adapter for hose connection incl. Pg 13.5 thread, platinum junction, electrolyte L 300, sphere membrane, S-glass, length 120 mm, 12 mm Ø, +10...+135°C, 0. 1 * 1 items
Catalog # SCOI285118200
Supplier:  SI Analytics
Frequently Bought Together