ContactUsLinkComponent
pH combination electrode with Memosens screw plug head (ATEX II 1/G) low-maintenance with NTC 30K, glass shaft, SILAMID reference, 2 hole junction, DuraLid reference system, sphere membrane, H glass, length 120 mm, 12 mm Ø, 0...14 pH, 0...130 °C 1 * 1 items
Catalog # SCOI285113495
Supplier:  SI Analytics
pH combination electrode with Memosens screw plug head (ATEX II 1/G) low-maintenance with NTC 30K, glass shaft, SILAMID reference, 2 hole junction, DuraLid reference system, sphere membrane, H glass, length 120 mm, 12 mm Ø, 0...14 pH, 0...130 °C 1 * 1 items
Catalog # SCOI285113495
Supplier:  SI Analytics
Frequently Bought Together