Agilent J and W HP-PLOT Al2O3 KCl PT, 50 m, 0.53 mm, 15 µm GC column, with two particle traps (front and rear) 1 * 1 items
Catalog # HEWL19095P-K25PT
Supplier: Agilent
Agilent J and W HP-PLOT Al2O3 KCl PT, 50 m, 0.53 mm, 15 µm GC column, with two particle traps (front and rear) 1 * 1 items
Catalog # HEWL19095P-K25PT
Supplier: Agilent
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection