ContactUsLinkComponent
UHPLC GRD, ECL. XDB-C18,2.1MM,1.8UM 1 * 3 items
Catalog # HEWL821725-903
Supplier:  Agilent
undefined
UHPLC GRD, ECL. XDB-C18,2.1MM,1.8UM 1 * 3 items
Catalog # HEWL821725-903
Supplier:  Agilent
Frequently Bought Together