Biochrom 30+ Amino Acid Analyser. Protein Hydrolysate system comprising instrument, 84 position cooled autosampler, Analytical column with top up resin, prewash column, BioSYS operating software, Ezchrom Elite data manipulation software, chemical kit 1 * 1 items
WAPR80-6000-61
Frequently Bought Together