ContactUsLinkComponent
Pendulum, simple study, dry and fluid damping, sensor without contact, the apparatus is composed of metallic structure, interface bo x with software, optical sensor (w/o friction), 5 sets of 4 weights (50, 100, 200, 500 and 1000g). 1 * 1 items
Catalog # DIDAPHD006580
Supplier:  DIDALAB
Pendulum, simple study, dry and fluid damping, sensor without contact, the apparatus is composed of metallic structure, interface bo x with software, optical sensor (w/o friction), 5 sets of 4 weights (50, 100, 200, 500 and 1000g). 1 * 1 items
Catalog # DIDAPHD006580
Supplier:  DIDALAB
Frequently Bought Together