ContactUsLinkComponent
Holes and young's slits, on a 40mm diameter glass plate, 6 single holes diamter 20, 30, 50, 100, 200, 500µm, 3 double holes diamter 70, distance 100, 200, 500µm, 2 squares 70x70µm, 200x200µm, 1 rectangle 70x200µm. 1 * 1 items
Catalog # DIDAPOD066730
Supplier:  DIDALAB
Holes and young's slits, on a 40mm diameter glass plate, 6 single holes diamter 20, 30, 50, 100, 200, 500µm, 3 double holes diamter 70, distance 100, 200, 500µm, 2 squares 70x70µm, 200x200µm, 1 rectangle 70x200µm. 1 * 1 items
Catalog # DIDAPOD066730
Supplier:  DIDALAB
Frequently Bought Together