ContactUsLinkComponent
Ruthenium Ru - 100 mg/l Standard for ICP/MS in HCl 1 * 100 mL
  ACSDM348.2CP.L1
 :  CPACHEM
Ruthenium Ru - 100 mg/l Standard for ICP/MS in HCl 1 * 100 mL
  ACSDM348.2CP.L1
 :  CPACHEM
Frequently Bought Together