Scandium Sc - 10 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM451.2NP.L1
Supplier: CPACHEM
Scandium Sc - 10 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM451.2NP.L1
Supplier: CPACHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Nitric acid 67%, NORMATOM®, Ultrapure for trace metal analysis
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ