Phosphorus P - 10 mg/l Standard for ICP/MS in H2O 1 * 100 mL
Catalog # ACSDM440.W.L1
Supplier: CPACHEM
Phosphorus P - 10 mg/l Standard for ICP/MS in H2O 1 * 100 mL
Catalog # ACSDM440.W.L1
Supplier: CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
UV/Visible standards, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals