ContactUsLinkComponent
SST 3-Port Assy, 5L - Stainless Steel Port Assemblies are designed for use with Media Storage Bottles for adding & withdrawing media . 3Port models feature siphon, fill and vent tubes and is supplied with a 45mm Polypropylene blue open top screw cap 1 * 1 items
Catalog # BELC5637-30005R
Supplier:  BELLCO GLASS
SST 3-Port Assy, 5L - Stainless Steel Port Assemblies are designed for use with Media Storage Bottles for adding & withdrawing media . 3Port models feature siphon, fill and vent tubes and is supplied with a 45mm Polypropylene blue open top screw cap 1 * 1 items
Catalog # BELC5637-30005R
Supplier:  BELLCO GLASS
Frequently Bought Together