ContactUsLinkComponent
SICCO Maxi 2-Vitrum-Desiccator, Antistatic. Size : Overall dimensions W x H x D mm 560 x 1150 x 580. Two compartments, two doors, usable with up to 17 shelves per compartment, standard is two shelves made of stainless steel, one tray and one hygromet 1 * 1 items
Catalog # BOHLV1845-07
Supplier:  Bohlender
undefined
SICCO Maxi 2-Vitrum-Desiccator, Antistatic. Size : Overall dimensions W x H x D mm 560 x 1150 x 580. Two compartments, two doors, usable with up to 17 shelves per compartment, standard is two shelves made of stainless steel, one tray and one hygromet 1 * 1 items
Catalog # BOHLV1845-07
Supplier:  Bohlender
Frequently Bought Together