ContactUsLinkComponent
Human CG018 full-length ORF ( NP_001073159.1, 1 A.A. - 173 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00090634-P012
Supplier:  Abnova
Human CG018 full-length ORF ( NP_001073159.1, 1 A.A. - 173 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00090634-P012
Supplier:  Abnova
Frequently Bought Together