Human TUBA8 partial ORF ( NP_061816.1, 1 A.A. - 89 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Katalogové číslo ABNOH00051807-Q012
Supplier: Abnova
Human TUBA8 partial ORF ( NP_061816.1, 1 A.A. - 89 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Katalogové číslo ABNOH00051807-Q012
Supplier: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK