Human PLXNA3 partial ORF ( NP_059984, 23 A.A. - 128 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
ABNOH00055558-Q012
: Abnova
Human PLXNA3 partial ORF ( NP_059984, 23 A.A. - 128 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
ABNOH00055558-Q012
: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection