ContactUsLinkComponent
Human B3GAT1 partial ORF (ABM86204.1, 28 A.A. - 334 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Katalogové číslo ABNOH00027087-Q022
Supplier:  Abnova
Human B3GAT1 partial ORF (ABM86204.1, 28 A.A. - 334 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Katalogové číslo ABNOH00027087-Q022
Supplier:  Abnova
Frequently Bought Together