ContactUsLinkComponent
Human DHRS2 partial ORF ( NP_878912.1, 229 A.A. - 300 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00010202-Q012
Supplier:  Abnova
Human DHRS2 partial ORF ( NP_878912.1, 229 A.A. - 300 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00010202-Q012
Supplier:  Abnova
Frequently Bought Together