ContactUsLinkComponent
Mphosph6 (human) recombinant protein (p01). human mphosph6 full-length orf ( aah05242, 1 a.a. - 160 a.a.) recombinant protein with g st-tag at n-terminal. 1 * 25 µG
Catalog # ABNOH00010200-P012
Supplier:  Abnova
Mphosph6 (human) recombinant protein (p01). human mphosph6 full-length orf ( aah05242, 1 a.a. - 160 a.a.) recombinant protein with g st-tag at n-terminal. 1 * 25 µG
Catalog # ABNOH00010200-P012
Supplier:  Abnova
Frequently Bought Together