Order Entry
Czech Republic
ContactUsLinkComponent
Calibration gas 25 h2s,50co,2.2%ch4,18%o2,n2 58l 4g18x22w25h50n2-58 1 * 1 items
Catalog # BWTE4G18X22W25H50
Supplier:  BW TECHNOLOGIES
Calibration gas 25 h2s,50co,2.2%ch4,18%o2,n2 58l 4g18x22w25h50n2-58 1 * 1 items
Catalog # BWTE4G18X22W25H50
Supplier:  BW TECHNOLOGIES
Frequently Bought Together