Human JAG2 partial ORF (NP_002217.3, 869 A.A. - 966 A.A.) Recombinant protein with GST TAG AT N-terminal. 1 * 25 µG
ABNOH00003714-Q022
: Abnova
Human JAG2 partial ORF (NP_002217.3, 869 A.A. - 966 A.A.) Recombinant protein with GST TAG AT N-terminal. 1 * 25 µG
ABNOH00003714-Q022
: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK