Order Entry
Czech Republic
ContactUsLinkComponent
Calibration gas 40 h2s,2%co2,2.5%ch4,15%o2,n2 34l 4gas15x25w40h2-34 1 * 1 items
Catalog # BWTE4GAS15X25W40H2
Supplier:  BW TECHNOLOGIES
Calibration gas 40 h2s,2%co2,2.5%ch4,15%o2,n2 34l 4gas15x25w40h2-34 1 * 1 items
Catalog # BWTE4GAS15X25W40H2
Supplier:  BW TECHNOLOGIES
Frequently Bought Together