ContactUsLinkComponent
Chromaster HPLC pack including 5160 600 Bar Manual Purge Pump with degassing unit and low pressure gradient included but not installed. 5260 Autosampler with Peltier, 5310 Column Oven, Organiser and interface control board. 1 * 1 items
Catalog # 903-0592
Supplier:  VWR Collection
Chromaster HPLC pack including 5160 600 Bar Manual Purge Pump with degassing unit and low pressure gradient included but not installed. 5260 Autosampler with Peltier, 5310 Column Oven, Organiser and interface control board. 1 * 1 items
Catalog # 903-0592
Supplier:  VWR Collection
Frequently Bought Together