ContactUsLinkComponent
LPTFE DIAPHRAGM VACUUM PUMP - INCLUDES ALL FITTINGS AND VACUUM HOSE MANIFOLD.20L/MINUTE PUMPING SPEED, ULTIMATE VACUUM - 6 TORR (220 V) 1 * 1 items
Catalog # IBSAPS-MD-VP-220
Supplier:  IBS Stock Items
LPTFE DIAPHRAGM VACUUM PUMP - INCLUDES ALL FITTINGS AND VACUUM HOSE MANIFOLD.20L/MINUTE PUMPING SPEED, ULTIMATE VACUUM - 6 TORR (220 V) 1 * 1 items
Catalog # IBSAPS-MD-VP-220
Supplier:  IBS Stock Items
Frequently Bought Together