Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Filtering flask

Filtering flask

Vacuum filtering flask, for 25 mm membranes, 125 ml
Catalog # GLAS259.245.04
Supplier:   GLASSCO
Filtering flask
Catalog # GLAS259.245.04
Supplier:  GLASSCO
Filtering flask
Filtering flask

Vacuum filtering flask, for 25 mm membranes, 125 ml

Price
Inquire For Price
Log in for availability
1 * 1 KS
Product Details & Documents

Flasks are made of heat resistant borosilicate glass 3.3, therefore they are suitable for hot and cold filtration.

  • Highly resistant against chemicals and thermal shock
  • High temperature resistance
  • With white inscription field

Due to the heavy wall thickness the VWR® filtration flask is vacuum tight.

Specifications
  • Capacity:
    125 ml
  • Description:
    Vacuum filtering flask, for 25 mm membranes
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.