ContactUsLinkComponent
Counting chamber, neubauer-improved (petroff type), depth 0.02mm, double net ruling, dark line, with clamps, with two cover glasses of 0.4mm thickness. 1 * 1 items
  MARI0642111
Counting chamber, neubauer-improved (petroff type), depth 0.02mm, double net ruling, dark line, with clamps, with two cover glasses of 0.4mm thickness. 1 * 1 items
  MARI0642111
Frequently Bought Together