Pipeta Pasteurova, 1 ml, bal. á 500 ks (nesterilní) 1 * 500 KS
Catalog # FLME27054
Supplier: FL MEDICAL
Pipeta Pasteurova, 1 ml, bal. á 500 ks (nesterilní) 1 * 500 KS
Catalog # FLME27054
Supplier: FL MEDICAL
Frequently Bought Together
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
Laboratory trays
Supplier: KARTELL
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
Centrifuge tubes, Oak Ridge, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
Cryostorage boxes, KeepIT-100™, WHEATON®
Supplier: DWK Life Sciences
Cryoboxes, for microtubes, B30/B32-series
Supplier: GLW
Chromatography papers
Supplier: Ahlstrom-Munksjö
Accessories for empty columns XK series
Supplier: Cytiva
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
CryoStorage vials
Supplier: EPPENDORF
Seals for ALPS™ 50 V heat sealer
Supplier: THERMO MATRIX TECHNOLOGIES
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with/without screw caps
Supplier: Thermo Fisher Scientific
384-well (HTS) microplates, non treated, BRANDplates®
Supplier: Brand