ContactUsLinkComponent
PLAIN DISC GL.MICROFIBRES FILT.1,0µM25MM 1 * 50 items
Catalog # FIOR0260A00001
Supplier:  Ahlstrom Munksjö
PLAIN DISC GL.MICROFIBRES FILT.1,0µM25MM 1 * 50 items
Catalog # FIOR0260A00001
Supplier:  Ahlstrom Munksjö
Frequently Bought Together